Ethyl acetoacetate 99+%, extra pure

Supplier: Thermo Scientific Chemicals

Synonyms: Acetoacetic acid ethyl ester, EAA

22040-0025 22040-1000 22040-5000
TS22040-0025EA 105 USD
TS22040-0025 TS22040-1000 TS22040-5000
Ethyl acetoacetate 99+%, extra pure
Ethyl acetoacetate
Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 180.8
Melting Pt: -45
Density: 1.03 g/cm³
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00009199
CAS Number: 141-97-9
EINECS: 205-516-1
Merck Index: 13,03793

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4180 to 1.4200 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥99.0 %
Color scale ≤10 APHA
Acidity (CH3COOH) ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR