Indigotrisulfonic acid potassium salt

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Potassium indigotrisulphonate

21517-0010 21517-0050 21517-0250
TS21517-0010EA 61 USD
TS21517-0010 TS21517-0050 TS21517-0250
Indigotrisulfonic acid potassium salt
Indigotrisulfonic acid potassium salt
Formula: C₁₆H₇K₃N₂O₁₁S₃
MW: 616.71 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00013160
CAS Number: 67627-18-3

Order Now

Specification Test Results

Appearance Dark blue Crystalline powder
Infrared spectrum Conforms
Molar extinction coefficient ≥ 17000 (250 - 256 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR