Stains-all 97%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Ethyl-2-[3-(1-ethylnaphtho[1,2-d]thiazolin-2-ylidene)-2-methylpropenyl]naphtho[1,2-d]thiazolium bromide, 3,3′-Diethyl-9-methyl-4,5,4′,5′-dibenzothiacarbocyanine
21351-0010
21351-0050
21351-2500
TS21351-0010EA
97
USD
TS21351-0010
TS21351-0050
TS21351-2500
Stains-all 97%, pure
Stains-all
Formula:
C₃₀H₂₇BrN₂S₂ MW: 559.584 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00078788 CAS Number: 7423-31-6 |
Specification Test Results
Appearance | Dark grey to dark purple Powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
TLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...