D-(-)-Diethyl tartrate (made from unnatural tartaric acid) 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (2S,3S)-Diethyl 2,3-dihydroxysuccinate, (S,S)-(-)-Diethyl tartrate, (2S,3S)-(-)-Diethyl tartrate, (-)-Diethyl D-tartrate, (S,S)-(-)-Tartaric acid diethyl ester, D(-)-Tartaric acid diethyl ester

20034-0250 20034-1000 20034-5000
TS20034-0050EA 30.9 USD
TS20034-0050 TS20034-0250 TS20034-1000 TS20034-5000
D-(-)-Diethyl tartrate (made from unnatural tartaric acid) 99%
D-(-)-Diethyl tartrate
Formula: C₈H₁₄O₆
Boiling Pt: 280 °C (760 mmHg)
Melting Pt: 17 °C
Density: 1.2 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00064451
CAS Number: 13811-71-7
EINECS: 237-458-8

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4470 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation -7.5° to -9° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR