Di-tert-butyl pyrocarbonate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Boc Anhydride, Di-tert-butyl dicarbonate, DIBOC
19467-0100
19467-0250
19467-0500
19467-1000
19467-5000
TS19467-0100EA
59
USD
TS19467-0100
TS19467-0250
TS19467-0500
TS19467-1000
TS19467-5000
Di-tert-butyl pyrocarbonate 99%
Di-tert-butyl pyrocarbonate
Formula:
C₁₀H₁₈O₅ MW: 218.25 g/mol Boiling Pt: 56…57 °C (.5 mmHg) Melting Pt: 22…24 °C Density: 1.02 g/cm³ Flash Pt: 37 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008805 CAS Number: 24424-99-5 EINECS: 246-240-1 UN: 2930 ADR: 6.1,II |
Specification Test Results
Appearance | White Low melting crystalline solid |
Infrared spectrum | Conforms |
Titration by coupling | 98.5 to 101.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...