Sudan orange G, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Solvent Orange 1, 4-(Phenylazo)resorcinol

19017-0010 19017-0050 19017-0250 19017-1000
TS19017-0010EA 1165 USD
TS19017-0010 TS19017-0050 TS19017-0250 TS19017-1000
Sudan orange G, pure
Sudan orange G
Formula: C₁₂H₁₀N₂O₂
Melting Pt: 143…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00002275
CAS Number: 2051-85-6
EINECS: 218-131-9

Order Now

Specification Test Results

Appearance Orange-red Powder
Infrared spectrum Conforms
UV spectrum Complies


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR