18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
18156-0050
18156-0250
18156-1000
18156-5000
TS18156-0050EA
48
USD
TS18156-0050
TS18156-0250
TS18156-1000
TS18156-5000
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) 99%
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane)
Formula:
C₁₂H₂₄O₆ Melting Pt: 36…40 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005113 CAS Number: 17455-13-9 EINECS: 241-473-5 Merck Index: 13,02630 |
Specification Test Results
Appearance | Clear colorless to white Crystals or crystalline mass or liquid |
Infrared spectrum | Conforms |
Melting point | 36 to 40 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...