Naphthalene 99%

Supplier: Thermo Scientific Chemicals
Warning

18090-0010 18090-0025 18090-0050 18090-2500
TS18090-0010EA 58 USD
TS18090-0010 TS18090-0025 TS18090-0050 TS18090-2500
Naphthalene 99%
Naphthalene
Formula: C₁₀H₈
Boiling Pt: 218 °C (760 mmHg)
Melting Pt: 79…82 °C
Density: 0.99 g/cm³
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00001742
CAS Number: 91-20-3
EINECS: 202-049-5
UN: 1334
ADR: 4.1,III
Merck Index: 12,06457
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c87c606cb828e0f39bab8b9d50c2c26501e1637d&vid=9e4bc21353014391fc8486208c8757e6375e2ef5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals or flakes or tablets or balls
Infrared spectrum Conforms
Melting point 79 to 82 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR