Benzeneselenenyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

17951-0010 17951-0100 17951-0250 17951-0500
TS17951-0010EA 31 USD
TS17951-0010 TS17951-0100 TS17951-0250 TS17951-0500
Benzeneselenenyl chloride 98%
Benzeneselenenyl chloride
Formula: (C₆H₅)SeCl
Boiling Pt: 120 °C (20 mmHg)
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00000478
CAS Number: 5707-04-0
EINECS: 227-196-2
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Orange to dark red Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 59 to 64 °C (Dec)
Titration Argentometric ≥97.5 %
Crosslinking 25 % DVB


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR