9-Fluorenylmethyl chloroformate (Fmoc chloride) 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (9H-Fluoren-9-yl)methyl carbonochloridate, 9-Fluorenylmethyl chloroformate, Fmoc-Cl, 9H-Fluoren-9-ylmethyl chloroformate, Chloroformic acid 9-fluorenylmethyl ester, FMOCCl

17094-0010 17094-0050 17094-0250 17094-1000
TS17094-0010EA 32 USD
TS17094-0010 TS17094-0050 TS17094-0250 TS17094-1000
9-Fluorenylmethyl chloroformate (Fmoc chloride) 98%
9-Fluorenylmethyl chloroformate (Fmoc chloride)
Formula: C₁₅H₁₁ClO₂
Melting Pt: 63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001138
CAS Number: 28920-43-6
EINECS: 249-313-6
UN: 2928
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 60 to 64 °C
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR