Acetylacetone 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: ACAC, 2,4-Pentanedione

12996-0010 12996-0025 12996-0050 12996-0100 12996-2500
TS12996-0010EA 82 USD
TS12996-0010 TS12996-0025 TS12996-0050 TS12996-0100 TS12996-2500
Acetylacetone 99+%
Acetylacetone
Formula: CH₃COCH₂COCH₃
MW: 100.12 g/mol
Boiling Pt: 140.4
Melting Pt: -23
Density: 0.975 g/cm³
Flash Pt: 35.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00008787
CAS Number: 123-54-6
EINECS: 204-634-0
UN: 2310
ADR: 3 III
Merck Index: 13,00083
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c85a64bf9c7997e9ab03e421a93e7c71e57b8a3&vid=134ed3029693deef3759692be256c57b3a58b087&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4510 to 1.4530 (20°C, 589 nm)
Water ≤0.1 %
GC ≥99.0 %
Color scale ≤50 APHA
Specific gravity (20°C) 0.972 to 0.978
Acidity (CH3COOH) ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR