(3-Carboxypropyl)triphenylphosphonium bromide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Carboxypropyltriphenylphosphonium bromide
33893-0250
33893-1000
TS33893-0250EA
63
USD
TS33893-0250
TS33893-1000
(3-Carboxypropyl)triphenylphosphonium bromide 97%
(3-Carboxypropyl)triphenylphosphonium bromide
Formula:
C₂₂H₂₂BrO₂P MW: 429.30 g/mol Melting Pt: 247…249 °C Storage Temperature: Ambient |
MDL Number:
MFCD00274196 CAS Number: 17857-14-6 |
Specification Test Results
Appearance | White Powder and/or granules |
Infrared spectrum | Conforms |
Melting point | 243 to 250 °C |
Titration Argentometric | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...