1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: EDC Methiodide, 3-(((Ethylimino)methylene)amino)-N,N,N-trimethylpropan-1-aminium iodide, 1-[3-(Dimethylamino)propyl]-3-ethylcarbodiimide Methiodide

33412-0010 33412-0100
TS33412-0010EA 31 USD
TS33412-0010 TS33412-0100
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide 98%
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide
Formula: CH₃CH₂N=C=N(CH₂)₃N(CH₃)₃I
MW: 297.17 g/mol
Melting Pt: 92…97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011833
CAS Number: 22572-40-3

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 92 to 97 °C
Titration Argentometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR