(Bromomethyl)triphenylphosphonium bromide 98%

Supplier: Thermo Scientific Chemicals
Warning

32148-0050 32148-0250
TS32148-0050EA 55 USD
TS32148-0050 TS32148-0250
(Bromomethyl)triphenylphosphonium bromide 98%
(Bromomethyl)triphenylphosphonium bromide
Formula: C₁₉H₁₇Br₂P
MW: 436.1200 g/mol
Melting Pt: 230…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00011864
CAS Number: 1034-49-7
EINECS: 213-857-2

Order Now

Specification Test Results

Appearance White to light beige to pink Powder or crystals
Infrared spectrum Conforms
Melting point 232 to 242 °C
Titration Argentometric ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR