Tris(2-aminoethyl)amine 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
31479-1000
31479-5000
TS31479-1000EA
223
USD
TS31479-1000
TS31479-5000
Tris(2-aminoethyl)amine 96%
Tris(2-aminoethyl)amine
Formula:
C₆H₁₈N₄ Boiling Pt: 265 °C (760 mmHg) Melting Pt: –16 °C Density: 0.977 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008177 CAS Number: 4097-89-6 EINECS: 223-857-4 UN: 2922 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow-green Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4960 to 1.4980 (20°C, 589 nm) |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...