1-Chloroethyl chloroformate 97+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Chloroethyl carbonochloridate

31446-0250 31446-1000
TS31446-0250EA 106 USD
TS31446-0250 TS31446-1000
1-Chloroethyl chloroformate 97+%
1-Chloroethyl chloroformate
Formula: C₃H₄Cl₂O₂
MW: 142.97 g/mol
Boiling Pt: 118…119 °C
Melting Pt: -65
Density: 1.32 g/cm³
Flash Pt: 40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000647
CAS Number: 50893-53-3
EINECS: 256-834-2
UN: 2742
ADR: 6.1 II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4240 (20°C, 589 nm)
GC ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR