Diphenylphosphine oxide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
31012-0010
31012-0050
TS31012-0010EA
56
USD
TS31012-0010
TS31012-0050
Diphenylphosphine oxide 97%
Diphenylphosphine oxide
Formula:
C₁₂H₁₁OP Melting Pt: 50…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002079 CAS Number: 4559-70-0 |
Specification Test Results
Appearance | White to yellow Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 50 to 57 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...