Tris(4-fluorophenyl)phosphine 98+%

Supplier: Thermo Scientific Chemicals
Warning

30634-0010 30634-0050
TS30634-0010EA 48 USD
TS30634-0010 TS30634-0050
Tris(4-fluorophenyl)phosphine 98+%
Tris(4-fluorophenyl)phosphine
Formula: C₁₈H₁₂F₃P
Melting Pt: 79…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00013553
CAS Number: 18437-78-0

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 79 to 83 °C
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR