Bromophenol red, pure

Supplier: Thermo Scientific Chemicals

30455-0050 30455-0250
TS30455-0050EA 104 USD
TS30455-0050 TS30455-0250
Bromophenol red, pure
Bromophenol red
Formula: C₁₉H₁₂Br₂O₅S
Melting Pt: 230 °C
MDL Number: MFCD00023014
CAS Number: 2800-80-8
EINECS: 220-538-1

Order Now

Specification Test Results

Appearance Red-brown to brown Powder
Infrared spectrum Conforms
Loss on drying ≤5 % (110°C)
Solubility (1 % in ethanol) Clear
Visual transition interval from pH 5.2 (yellow-red) to pH 7.0 (purple)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR