N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate, TSTU, 2-(2,5-Dioxopyrrolidin-1-yl)-1,1,3,3-tetramethylisouronium tetrafluoroborate
30319-0010
30319-0050
TS30319-0010EA
72
USD
TS30319-0010
TS30319-0050
N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate 97%
N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate
Formula:
C₉H₁₆BF₄N₃O₃ Melting Pt: 194…200 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00077875 CAS Number: 105832-38-0 |
Specification Test Results
Appearance | White to off-white Crystalline powder or needles |
Infrared spectrum | Conforms |
Elemental Analysis | 13.40 to 14.52 % (N) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...