Tetrakis(1-pyrazolyl)borate potassium salt 95%
Supplier: Thermo Scientific Chemicals
30016-0010
30016-0050
TS30016-0010EA
107
USD
TS30016-0010
TS30016-0050
Tetrakis(1-pyrazolyl)borate potassium salt 95%
Tetrakis(1-pyrazolyl)borate potassium salt
Formula:
C₁₂H₁₂BKN₈O₄ Melting Pt: 253…254 °C Storage Temperature: Ambient |
MDL Number:
MFCD00800557 CAS Number: 14782-58-2 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...