(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
29706-0010
29706-0100
TS29706-0010EA
27
USD
TS29706-0010
TS29706-0100
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one 98%
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one
Formula:
C₆H₇NO Boiling Pt: 102…106 °C (.2hPa) Melting Pt: 55…59 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00213364 CAS Number: 49805-30-3 EINECS: 421-830-3 |
Specification Test Results
Appearance | Off-white to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 55 to 59 °C |
GC | ≥97.5 % |
UV | at 400 nm A: ≤2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...