1,4-Bis(diphenylphosphino)butane 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: DPPB, 1,4-Bis(diphenylphosphino)butane, Butylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)butane, Butane-1,3-diylbis(diphenylphosphine)

29646-0050 29646-0250
TS29646-0050EA 50 USD
TS29646-0050 TS29646-0250
1,4-Bis(diphenylphosphino)butane 98%
1,4-Bis(diphenylphosphino)butane
Formula: C₂₈H₂₈P₂
Melting Pt: 130…137 °C
MDL Number: MFCD00003051
CAS Number: 7688-25-7
EINECS: 231-698-7

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 132 to 137 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR