(Chloromethylene)dimethylammonium chloride 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-Chloromethylen-N,N-Dimethyl Ammonium Chloride
29577-0050
29577-0250
TS29577-0050EA
43
USD
TS29577-0050
TS29577-0250
(Chloromethylene)dimethylammonium chloride 96%
(Chloromethylene)dimethylammonium chloride
Formula:
C₃H₇Cl₂N MW: 128.00 g/mol Melting Pt: 139 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00011868 CAS Number: 3724-43-4 EINECS: 425-970-6 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | Light yellow to light brown Crystalline powder |
Infrared spectrum | Conforms |
Titration Argentometric | 95 to 105 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...