Bis(diphenylphosphino)methane 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: [(1,1-Diphenylphosphino)methyl](diphenyl)phosphine, DPPM

29361-0050 29361-0250
TS29361-0050EA 68 USD
TS29361-0050 TS29361-0250
Bis(diphenylphosphino)methane 98%
Bis(diphenylphosphino)methane
Formula: C₂₅H₂₂P₂
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00003537
CAS Number: 2071-20-7
EINECS: 218-194-2

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 118 to 122 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR