4,4-Dimethoxybutanone 92%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4,4-Dimethoxybutan-2-one, 3-Oxobutyraldehyde dimethyl acetal, Acetoacetaldehyde-1,1-Dimethyl Acetal, Acetylacetaldehyde dimethyl acetal, 4,4-Dimethoxy-2-butanone
29233-0010
29233-2500
TS29233-0010EA
296
USD
TS29233-0010
TS29233-2500
4,4-Dimethoxybutanone 92%
4,4-Dimethoxybutanone
Formula:
C₆H₁₂O₃ MW: 132.16 g/mol Boiling Pt: 178 °C (1013 hPa) Melting Pt: –82 °C Density: 0.99 g/cm³ Flash Pt: 49 °C |
MDL Number:
MFCD00008789 CAS Number: 5436-21-5 EINECS: 226-605-1 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4130 to 1.4160 (20°C, 589 nm) |
GC | ≥90 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...