Calix[4]arene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3,5,7(1,3)-Tetrabenzenacyclooctaphane-12,32,52,72-tetraol

29019-0010 29019-0050
TS29019-0010EA 159 USD
TS29019-0010 TS29019-0050
Calix[4]arene 98%
Calix[4]arene
Formula: C₂₈H₂₄O₄
MW: 424.5 g/mol
Melting Pt: 315…318 °C
Storage Temperature: Ambient
MDL Number: MFCD00233673
CAS Number: 74568-07-3

Order Now

Specification Test Results

Appearance Off-white to light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR