Ruthenium(III) chloride oxide ≥85%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ruthenium oxychloride, Tetraamminechlorohydroxoruthenium(III) chloride, Ruthenium Red

27971-0010 27971-0050
TS27971-0010EA 129 USD
TS27971-0010 TS27971-0050
Ruthenium(III) chloride oxide ≥85%, pure
Ruthenium(III) chloride oxide
Formula: H₄₂Cl₆N₁₄O₂Ru₃
MW: 786.354 g/mol
Melting Pt: >500 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011479
CAS Number: 11103-72-3

Order Now

Specification Test Results

Appearance Dark red to brown to purple Crystalline powder
Elemental Analysis 34 to 39 % (Ru)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR