Fehling's reagent I (copper(II) sulphate solution)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Fehling A solution

27080-0010 27080-0025
TS27080-0010EA 53 USD
TS27080-0010 TS27080-0025
Fehling's reagent I (copper(II) sulphate solution)
Fehling's reagent I (copper(II) sulphate solution)
Formula: CuSO₄
MW: 159.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear blue Solution
Solution contains 69.2 g CuSO4.5H2O/l H2O

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR