(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) 99%

Supplier: Thermo Scientific Chemicals
Warning

27072-0050 27072-0250
TS27072-0050EA 89 USD
TS27072-0050 TS27072-0250
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) 99%
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
Formula: C₁₈H₂₈F₆N₆OP₂
Melting Pt: 145…160 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077411
CAS Number: 128625-52-5

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 145 to 160 °C (Dec)
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR