(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(-)-MTPA-Cl, (-)-Mosher's Acid Chloride, (R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
26412-0010
26412-2500
TS26412-0010EA
449
USD
TS26412-0010
TS26412-2500
(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride 98+%
(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
Formula:
C₁₀H₈ClF₃O₂ MW: 252.62 g/mol Boiling Pt: 213…214 °C (760 mmHg) Density: 1.34 g/cm³ Flash Pt: 89 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00044400 CAS Number: 39637-99-5 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4675 to 1.4715 (20°C, 589 nm) |
GC | ≥98.0 % |
Specific optical rotation | -128° to -134° (20°C, 589 nm) (c=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...