Dimethyl (+)-2,3-O-isopropylidene-D-tartrate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
(+)-Dimethyl 2,3-O-isopropylidene-D-tartrate, Dimethyl (4S,5S)-2,2-Dimethyl-1,3-dioxolane-4,5-dicarboxylate, (4S,5S)-2,2-Dimethyl-1,3-dioxolane-4,5-dicarboxylic Acid Dimethyl Ester, (+)-2,3-O-Isopropylidene-D-tartaric Acid Dimethyl Ester
25844-0100
25844-0500
TS25844-0100EA
126
USD
TS25844-0100
TS25844-0500
Dimethyl (+)-2,3-O-isopropylidene-D-tartrate 97%
Dimethyl (+)-2,3-O-isopropylidene-D-tartrate
Formula:
C₉H₁₄O₆ MW: 218.21 g/mol Boiling Pt: 139 °C (21hPa) Density: 1.15 g/cm³ Flash Pt: 135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040504 CAS Number: 37031-30-4 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4386 to 1.4406 (20°C, 589 nm) |
GC | ≥96.0 % |
Specific optical rotation | +52° to +56° (20°C, 589 nm) (neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...