Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane) 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzo-18-crown 6-Ether

24398-0010 24398-0050
TS24398-0010EA 55 USD
TS24398-0010 TS24398-0050
Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane) 97%
Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane)
Formula: C₁₆H₂₄O₆
MW: 312.36 g/mol
Melting Pt: 40…45 °C
Storage Temperature: Ambient
MDL Number: MFCD00062741
CAS Number: 14098-24-9

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 40 to 45 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR