Trichloromethyl chloroformate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
24331-0100
24331-1000
TS24331-0100EA
80
USD
TS24331-0100
TS24331-1000
Trichloromethyl chloroformate 99%
Trichloromethyl chloroformate
Formula:
C₂Cl₄O₂ Boiling Pt: 128 °C (760 mmHg) Melting Pt: –57 °C Density: 1.64 g/cm³ Storage Temperature: Refrigerator |
MDL Number:
MFCD00015553 CAS Number: 503-38-8 EINECS: 207-965-9 UN: 3390 ADR: 6.1, Merck Index: 1303368 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4560 to 1.4590 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...