2,6-Di-O-methyl-β-cyclodextrine 96%

Supplier: Thermo Scientific Chemicals

Synonyms: Dimethyl-β-cyclodextrin, methylated β-cyclodextrins, Heptakis(2,6-di-o-methyl)-β-cyclodextrin, 2,6-Di-O-methyl-β-cyclodextrin

23553-0010 23553-0050
TS23553-0010EA 101 USD
TS23553-0010 TS23553-0050
2,6-Di-O-methyl-β-cyclodextrine 96%
2,6-Di-O-methyl-β-cyclodextrine
Formula: C₅₆H₉₈O₃₅
MW: 1331.3563 g/mol
Melting Pt: 298…310 °C
Storage Temperature: Ambient
MDL Number: MFCD00011616
CAS Number: 51166-71-3

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Assay ≥95 % Methylated beta-cyclodextrins ((on dry basis) calculated)
Water ≤5.0 % (K.F.)
HPLC 35.0 to 55.0 % (ELSD) isomer purity Heptakis(2,6-di-O-methyl)-beta-cyclodextrin
TLC ≤0.5 % unmodified beta-cyclodextrin
NMR 12.0 to 16.0 methyl groups per cyclodextrin ring (average degree of substitution)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR