Orange G 85+%, pure, high purity biological stain

Supplier: Thermo Scientific Chemicals

22982-0250 22982-1000
TS22982-0250EA 55 USD
TS22982-0250 TS22982-1000
Orange G 85+%, pure, high purity biological stain
Orange G
Formula: C₁₆H₁₀N₂Na₂O₇S₂
Melting Pt: 141 °C
Storage Temperature: Ambient
MDL Number: MFCD00012457
CAS Number: 1936-15-8
EINECS: 217-705-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR