Neutral red, pure, high purity biological stain
Supplier: Thermo Scientific Chemicals
Synonyms:
N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5
22981-0250
22981-1000
TS22981-0250EA
84
USD
TS22981-0250
TS22981-1000
Neutral red, pure, high purity biological stain
Neutral red
Formula:
C₁₅H₁₇ClN₄ MW: 288.78 g/mol Melting Pt: 290 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012651 CAS Number: 553-24-2 EINECS: 209-035-8 Merck Index: 1206571 |
Specification Test Results
Appearance | Dark green Crystalline powder |
Infrared spectrum | Conforms |
Dye content | ≥50 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...