Aniline blue, high purity biological stain

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: C.I. 42780

22966-0250 22966-1000
TS22966-0250EA 91 USD
TS22966-0250 TS22966-1000
Aniline blue, high purity biological stain
Methyl blue
Formula: C₃₇H₂₇N₃Na₂O₉S₃
Storage Temperature: Ambient
MDL Number: MFCD00058509
CAS Number: 28983-56-4
EINECS: 249-113-9

Order Now

Specification Test Results

Appearance Dark brown Glistening crystalline powder or crystals
Infrared spectrum Conforms
Water ≤10 %
UV Lambda max 594 to 610 nm (in 0.005M HCl)
Absorption Ratio P-15/P+15 0.96 to 1.09


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR