Crystal violet, pure, high purity biological stain
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
CI 42555
22964-1000
22964-5000
TS22964-1000EA
113
USD
TS22964-1000
TS22964-5000
Crystal violet, pure, high purity biological stain
Crystal violet
Formula:
C₂₅H₃₀ClN₃ MW: 407.99 g/mol Melting Pt: 173 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011750 CAS Number: 548-62-9 EINECS: 208-953-6 UN: 3077 ADR: 9 III Merck Index: 13,04406 |
Specification Test Results
Appearance | Dark green to green-brown Powder or crystals |
Infrared spectrum | Conforms |
Loss on drying | ≤9 % (110°C) |
Sulfated ash | ≤3 % |
Titration | ≥96.0 % (on dried substance) |
Lead (Pb) | ≤30 ppm |
Zinc (Zn) | Passes test |
Insoluble matter | (in ethanol) Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...