Cresyl violet (acetate), pure, high purity biological stain

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cresyl echt violet, Cresyl violet, Cresyl fast violet, 5,9-Diaminobenzo[a]phenoxazin-7-ium acetate, 9-Amino-5-imino-5H-benzo[a]phenoxazine acetate salt, Cresyl Violet acetate, Gentian violet

22963-0050 22963-0250
TS22963-0050EA 116 USD
TS22963-0050 TS22963-0250
Cresyl violet (acetate), pure, high purity biological stain
Cresyl violet (acetate)
Formula: C₁₈H₁₅N₃O₃
MW: 321.33 g/mol
Melting Pt: 140…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00013151
CAS Number: 10510-54-0

Order Now

Specification Test Results

Appearance Dark green to black Crystalline powder
Infrared spectrum Conforms
Dye content ≥65 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR