Brilliant cresyl blue, pure, high purity biological stain

Supplier: Thermo Scientific Chemicals

Synonyms: CI 51010, 1,3-Diamino-7-(diethylamino)-3-methylphenoxazin-5-ium tetrachlorozincate (2:1), Brilliant cresyl blue zinc chloride double salt, Brilliant Blue C, BCB

22959-0100 22959-0250
TS22959-0100EA 107 USD
TS22959-0100 TS22959-0250
Brilliant cresyl blue, pure, high purity biological stain
Brilliant cresyl blue
Formula: C₃₄H₄₂Cl₄N₈O₂Zn
MDL Number: MFCD00148901
CAS Number: 81029-05-2
EINECS: 279-675-0

Order Now

Specification Test Results

Appearance Green to dark blue Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤8 % (1 g, 110°C)
Solubility (0.1% in 50% ethanol) Clear blue
Specific absorbance A (1%/1cm) ≥1500 (625 to 630 nm, 50% ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR