Sodium diphenylamine-4-sulphonate, pure indicator grade, redox indicator

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium Diphenylaminesulphonate, Sodium N-phenylsulphanilate, Sodium (4-anilinobenzenesulphonate), Diphenylamine-4-sulphonic acid, sodium salt

22222-0050 22222-0250
TS22222-0050EA 115 USD
TS22222-0050 TS22222-0250
Sodium diphenylamine-4-sulphonate, pure indicator grade, redox indicator
Sodium diphenylamine-4-sulphonate
Formula: C₁₂H₁₀NNaO₃S
Storage Temperature: Ambient
MDL Number: MFCD00036468
CAS Number: 6152-67-6
EINECS: 228-165-6
Merck Index: 1307397

Order Now

Specification Test Results

Appearance Off-white to beige to light grey Crystalline powder
Infrared spectrum Conforms
Sensitivity (as indicator) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR