Phenyl chloroformate 99%

Supplier: Thermo Scientific Chemicals
Danger

21821-0010 21821-2500
TS21821-0010EA 277 USD
TS21821-0010 TS21821-2500
Phenyl chloroformate 99%
Phenyl chloroformate
Formula: C₇H₅ClO₂
Boiling Pt: 188…189 °C (1013mbar)
Melting Pt: –28 °C
Density: 1.244 g/cm³
Flash Pt: 80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000637
CAS Number: 1885-14-9
EINECS: 217-547-8
UN: 2746
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5100 to 1.5120 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤50 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR