Chlormequat chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chlorocholine chloride, 2-Chloroethyltrimethylammonium chloride, (2-Chloroethyl)trimethylammonium chloride

21542-0250 21542-1000
TS21542-0250EA 62 USD
TS21542-0250 TS21542-1000
Chlormequat chloride 98%
Chlormequat chloride
Formula: ClCH₂CH₂N(CH₃)₃Cl
MW: 158.07 g/mol
Melting Pt: 239…243 °C
Storage Temperature: Ambient
MDL Number: MFCD00011869
CAS Number: 999-81-5
EINECS: 213-666-4
UN: 2811
ADR: 6.1,III
Merck Index: 1302120

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤1 % (K.F.)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR