Indigo (synthetic)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pigment Blue 66, Vat Blue 1, Indigotin, 2,2'-Bis(2,3-dihydro-3-oxoindolyliden), Indigo (synthetic), 2-(1,3-Dihydro-3-oxo-2H-indol-2-ylidene)-1,2-dihydro-3H-indol-3-one

21213-0250 21213-1000
TS21213-0250EA 42 USD
TS21213-0250 TS21213-1000
Indigo (synthetic)
Indigo
Formula: C₁₆H₁₀N₂O₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00005722
CAS Number: 482-89-3
EINECS: 207-586-9
Merck Index: 1204977

Order Now

Specification Test Results

Appearance Dark blue to violet Powder
Infrared spectrum Conforms
UV spectrum Complies


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR