4-(Phenylazo)diphenylamine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Phenylazodiphenylamine
21201-0050
21201-0250
TS21201-0050EA
37
USD
TS21201-0050
TS21201-0250
4-(Phenylazo)diphenylamine 97%
4-(Phenylazo)diphenylamine
Formula:
C₁₈H₁₅N₃ MW: 273.34 g/mol Melting Pt: 84…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003023 CAS Number: 101-75-7 EINECS: 202-972-3 |
Specification Test Results
Appearance | Orange to brown Powder |
Infrared spectrum | Conforms |
Melting point | 89 to 91 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...