D-(-)-Diisopropyl tartrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (2S,3S)-(-)-Diisopropyl tartrate, (2S,3S)-Diisopropyl 2,3-dihydroxysuccinate, D-(-)-Tartaric acid diisopropyl ester

21118-0100 21118-0500
TS21118-0100EA 32 USD
TS21118-0100 TS21118-0500
D-(-)-Diisopropyl tartrate 99%
D(-)-Diisopropyl tartrate
Formula: C₁₀H₁₈O₆
MW: 234.25 g/mol
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008876
CAS Number: 62961-64-2
EINECS: 263-771-4

Order Now

Specification Test Results

Appearance Clear colorless Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4400 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation -16° to -18.5° (20°C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR