Benzyltrimethylammonium chloride 50% (w/w) in water

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N,N,N-Trimethyl-1-phenylmethanaminium chloride

20938-0010 20938-1000
TS20938-0010EA 116 USD
TS20938-0010 TS20938-1000
Benzyltrimethylammonium chloride 50% (w/w) in water
Benzyltrimethylammonium chloride
Formula: C₁₀H₁₆ClN
MW: 185.7 g/mol
Density: 1.044 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011782
CAS Number: 56-93-9
EINECS: 200-300-3
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Titration Argentometric ≥49 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR