Leuco crystal violet 99%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,4',4''-Methanetriyltris(N,N-dimethylaniline), Tris(4-dimethylaminophenyl)methane, 4,4',4''-Methylidynetris(N,N-dimethylaniline)

20433-0050 20433-0250
TS20433-0050EA 31 USD
TS20433-0050 TS20433-0250
Leuco crystal violet 99%, pure
Leuco crystal violet
Formula: C₂₅H₃₁N₃
MW: 373.54 g/mol
Melting Pt: 175…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00008314
CAS Number: 603-48-5
UN: 3077

Order Now

Specification Test Results

Appearance White to light grey to light violet Powder
Infrared spectrum Conforms
Melting point 175 to 180 °C
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR