Calcon® (mordant black 17), pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Palatine chrome black 6BN, Mordant Black 17, Eriochrome® Blue Black R, Sodium 3-hydroxy-4-((2-hydroxynaphthalen-1-yl)diazenyl)naphthalene-1-sulphonate, Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-1-naphthalenesulphonate

20244-0250 20244-1000
TS20244-0250EA 35 USD
TS20244-0250 TS20244-1000
Calcon® (mordant black 17), pure
Calcon® (mordant black 17)
Formula: C₂₀H₁₃N₂NaO₅S
Storage Temperature: Ambient
MDL Number: MFCD00004069
CAS Number: 2538-85-4
EINECS: 219-810-2

Order Now

Specification Test Results

Appearance Dark brown to black Powder
Infrared spectrum Conforms
UV spectrum Complies


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR